missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluorophenyl Acrylate (stabilized with MEHQ) 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 197°C |
| Quantity | 1 g |
| Formula Weight | 238.11 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Pentafluorophenyl Acrylate (stabilized with MEHQ) |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |