accessibility menu, dialog, popup

UserName

H2COOFFFFF

CAS RN 71195-85-2

IUPAC Name: 2,3,4,5,6-pentafluorophenyl prop-2-enoate
Synonyms: 2,3,4,5,6-pentafluorophenyl prop-2-enoate
Molecular Weight (g/mol): 238.11
Molecular Formula: C9H3F5O2
InChi Key: RFOWDPMCXHVGET-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C=C)C(F)=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results