missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 4-Nitrobenzenesulfonate 98.0+%, TCI America™

Catalog No. P22311G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22311G 1 g
P22315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22311G Supplier TCI America Supplier No. P22311G

Chemical Identifiers

CAS 244633-31-6
Molecular Formula C12H4F5NO5S
Molecular Weight (g/mol) 369.22
MDL Number MFCD05975120
InChI Key DQKVZFMQPLLPMR-UHFFFAOYSA-N
Synonym 4-Nitrobenzenesulfonic Acid Pentafluorophenyl Ester
PubChem CID 2783096
IUPAC Name 2,3,4,5,6-pentafluorophenyl 4-nitrobenzene-1-sulfonate
SMILES [O-][N+](=O)C1=CC=C(C=C1)S(=O)(=O)OC1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Melting Point 108°C
Color White-Yellow
Quantity 1 g
Formula Weight 369.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pentafluorophenyl 4-Nitrobenzenesulfonate