accessibility menu, dialog, popup

UserName

NOOSOOOFFFFF

CAS RN 244633-31-6

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 4-nitrobenzene-1-sulfonate
Synonyms: 2,3,4,5,6-pentafluorophenyl 4-nitrobenzenesulfonate
Molecular Weight (g/mol): 369.22
Molecular Formula: C12H4F5NO5S
InChi Key: DQKVZFMQPLLPMR-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)S(=O)(=O)OC1=C(F)C(F)=C(F)C(F)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results