missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluoroiodobenzene (stabilized with Copper chip) 99.0+%, TCI America™
$87.60 - $277.75
Chemical Identifiers
| CAS | 827-15-6 |
|---|---|
| Molecular Formula | C6F5I |
| Molecular Weight (g/mol) | 293.962 |
| MDL Number | MFCD00001032 |
| InChI Key | OPYHNLNYCRZOGY-UHFFFAOYSA-N |
| Synonym | iodopentafluorobenzene, pentafluoroiodobenzene, benzene, pentafluoroiodo, 2,3,4,5,6-pentafluoroiodobenzene, iodoperfluorobenzene, pentafluorophenyl iodide, 1,2,3,4,5-pentafluoro-6-iodo-benzene, perfluoroiodobenzene, ibf, pubchem19982 |
| PubChem CID | 70008 |
| IUPAC Name | 1,2,3,4,5-pentafluoro-6-iodobenzene |
| SMILES | C1(=C(C(=C(C(=C1F)F)I)F)F)F |
Chemical Identifiers
| 827-15-6 | |
| 293.962 | |
| OPYHNLNYCRZOGY-UHFFFAOYSA-N | |
| 70008 | |
| C1(=C(C(=C(C(=C1F)F)I)F)F)F |
| C6F5I | |
| MFCD00001032 | |
| iodopentafluorobenzene, pentafluoroiodobenzene, benzene, pentafluoroiodo, 2,3,4,5,6-pentafluoroiodobenzene, iodoperfluorobenzene, pentafluorophenyl iodide, 1,2,3,4,5-pentafluoro-6-iodo-benzene, perfluoroiodobenzene, ibf, pubchem19982 | |
| 1,2,3,4,5-pentafluoro-6-iodobenzene |
Specifications
| 827-15-6 | |
| 162°C | |
| MFCD00001032 | |
| iodopentafluorobenzene, pentafluoroiodobenzene, benzene, pentafluoroiodo, 2,3,4,5,6-pentafluoroiodobenzene, iodoperfluorobenzene, pentafluorophenyl iodide, 1,2,3,4,5-pentafluoro-6-iodo-benzene, perfluoroiodobenzene, ibf, pubchem19982 | |
| C1(=C(C(=C(C(=C1F)F)I)F)F)F | |
| 293.962 | |
| 293.96 | |
| Liquid |
| Yellow | |
| C6F5I | |
| 5 g | |
| OPYHNLNYCRZOGY-UHFFFAOYSA-N | |
| 1,2,3,4,5-pentafluoro-6-iodobenzene | |
| 70008 | |
| ≥99.0% (GC) | |
| Pentafluoroiodobenzene (stabilized with Copper chip) |
Safety and Handling
TSCA : No