missing translation for 'onlineSavingsMsg'
Learn More

Pentafluoroaniline, 97%

Catalog No. aaa1934822
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
5 g
Catalog No. Quantity
AAA1934822 100 g
AAA1934806 5 g
AAA1934814 25 g
3 options

Catalog No. AAA1934822

Supplier: Thermo Scientific Chemicals A1934822

Only null left

CAS: 771-60-8 | C6H2F5N | 183.08 g/mol

2,3,4,5,6-Pentafluoroaniline finds it uses in the preparation of pentafluorophenylammonium triflate, an efficient catalyst for esterification and thioesterification. It is also used in the synthesis of various titanium complexes having two anionic [N, O-] bidentate salicylaldiminato ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,4,5,6-Pentafluoroaniline finds it uses in the preparation of pentafluorophenylammonium triflate, an efficient catalyst for esterification and thioesterification. It is also used in the synthesis of various titanium complexes having two anionic [N, O-] bidentate salicylaldiminato ligands.

Solubility
Soluble in toluene, and organic solvents. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 771-60-8
Molecular Formula C6H2F5N
Molecular Weight (g/mol) 183.08
MDL Number MFCD00007643
InChI Key NOXLGCOSAFGMDV-UHFFFAOYSA-N
Synonym pentafluoroaniline, pentafluorophenylamine, aminopentafluorobenzene, benzenamine, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluorobenzenamine, aniline, 2,3,4,5,6-pentafluoro, unii-ms06il3y2q, ms06il3y2q, 2,3,4,5,6-pentafluorophenylamine, pubchem1546
PubChem CID 13040
SMILES NC1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Melting Point 34°C to 37°C
Density 1.744
Boiling Point 156°C to 157°C
Flash Point 73°C (163°F)
Quantity 100 g
Beilstein 1819387
Solubility Information Soluble in toluene,and organic solvents. Insoluble in water.
IUPAC Name pentafluoroaniline
Formula Weight 183.08
Percent Purity ≥98%
Chemical Name or Material Pentafluoroaniline
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 212-234-2
RTECSNumber BY7920000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only