accessibility menu, dialog, popup

UserName

NH2FFFFF

CAS RN 771-60-8

IUPAC Name: pentafluoroaniline
Synonyms: 2,3,4,5,6-pentafluoroaniline
Molecular Weight (g/mol): 183.08
Molecular Formula: C6H2F5N
InChi Key: NOXLGCOSAFGMDV-UHFFFAOYSA-N
SMILES: NC1=C(F)C(F)=C(F)C(F)=C1F
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)