missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluoroaniline, 97%
Description
2,3,4,5,6-Pentafluoroaniline finds it uses in the preparation of pentafluorophenylammonium triflate, an efficient catalyst for esterification and thioesterification. It is also used in the synthesis of various titanium complexes having two anionic [N, O-] bidentate salicylaldiminato ligands.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3,4,5,6-Pentafluoroaniline finds it uses in the preparation of pentafluorophenylammonium triflate, an efficient catalyst for esterification and thioesterification. It is also used in the synthesis of various titanium complexes having two anionic [N, O-] bidentate salicylaldiminato ligands.
Solubility
Soluble in toluene, and organic solvents. Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
2,3,4,5,6-Pentafluoroaniline finds it uses in the preparation of pentafluorophenylammonium triflate, an efficient catalyst for esterification and thioesterification. It is also used in the synthesis of various titanium complexes having two anionic [N, O-] bidentate salicylaldiminato ligands.
Solubility
Soluble in toluene, and organic solvents. Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 34°C to 37°C |
| Density | 1.744 |
| Boiling Point | 156°C to 157°C |
| Flash Point | 73°C (163°F) |
| Quantity | 25 g |
| Beilstein | 1819387 |
| Solubility Information | Soluble in toluene,and organic solvents. Insoluble in water. |
| IUPAC Name | 2,3,4,5,6-pentafluoroaniline |
| Formula Weight | 183.08 |
| Percent Purity | ≥98% |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H301-H315-H319-H335 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 212-234-2 |
| RTECSNumber | BY7920000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only