missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentaerythrityl Tetrachloride 98.0+%, TCI America™
$139.81 - $411.95
Chemical Identifiers
| CAS | 3228-99-7 |
|---|---|
| Molecular Formula | C5H8Cl4 |
| Molecular Weight (g/mol) | 209.92 |
| MDL Number | MFCD00000942 |
| InChI Key | KPZGRMZPZLOPBS-UHFFFAOYSA-N |
| Synonym | pentaerythrityl tetrachloride, 1,3-dichloro-2,2-bis chloromethyl propane, tetrachloride, tetrakis chloromethyl methane, pentaerythritol tetrachloride, methane, tetrakis chloromethyl, propane, 1,3-dichloro-2,2-bis chloromethyl, ccris 4944, 2,2-bis chloromethyl-1,3-dichloropropane |
| PubChem CID | 76700 |
| IUPAC Name | 1,3-dichloro-2,2-bis(chloromethyl)propane |
| SMILES | ClCC(CCl)(CCl)CCl |
Chemical Identifiers
| 3228-99-7 | |
| 209.92 | |
| KPZGRMZPZLOPBS-UHFFFAOYSA-N | |
| 76700 | |
| ClCC(CCl)(CCl)CCl |
| C5H8Cl4 | |
| MFCD00000942 | |
| pentaerythrityl tetrachloride, 1,3-dichloro-2,2-bis chloromethyl propane, tetrachloride, tetrakis chloromethyl methane, pentaerythritol tetrachloride, methane, tetrakis chloromethyl, propane, 1,3-dichloro-2,2-bis chloromethyl, ccris 4944, 2,2-bis chloromethyl-1,3-dichloropropane | |
| 1,3-dichloro-2,2-bis(chloromethyl)propane |
Specifications
| 3228-99-7 | |
| White-Yellow | |
| C5H8Cl4 | |
| 5 g | |
| KPZGRMZPZLOPBS-UHFFFAOYSA-N | |
| 1,3-dichloro-2,2-bis(chloromethyl)propane | |
| 76700 | |
| ≥98.0% (GC) | |
| Pentaerythrityl Tetrachloride |
| 96°C | |
| 110°C | |
| MFCD00000942 | |
| pentaerythrityl tetrachloride, 1,3-dichloro-2,2-bis chloromethyl propane, tetrachloride, tetrakis chloromethyl methane, pentaerythritol tetrachloride, methane, tetrakis chloromethyl, propane, 1,3-dichloro-2,2-bis chloromethyl, ccris 4944, 2,2-bis chloromethyl-1,3-dichloropropane | |
| ClCC(CCl)(CCl)CCl | |
| 209.92 | |
| 209.92 | |
| Crystalline Powder |
Safety and Handling
TSCA : No