missing translation for 'onlineSavingsMsg'
Learn More

Pentaerythrityl Tetrachloride 98.0+%, TCI America™

Catalog No. p129525g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
P129525G 25 g
P12955G 5 g
2 options

Catalog No. P129525G

Supplier: TCI America P129525G

Only null left

Chemical Identifiers

CAS 3228-99-7
Molecular Formula C5H8Cl4
Molecular Weight (g/mol) 209.92
MDL Number MFCD00000942
InChI Key KPZGRMZPZLOPBS-UHFFFAOYSA-N
Synonym pentaerythrityl tetrachloride, 1,3-dichloro-2,2-bis chloromethyl propane, tetrachloride, tetrakis chloromethyl methane, pentaerythritol tetrachloride, methane, tetrakis chloromethyl, propane, 1,3-dichloro-2,2-bis chloromethyl, ccris 4944, 2,2-bis chloromethyl-1,3-dichloropropane
PubChem CID 76700
IUPAC Name 1,3-dichloro-2,2-bis(chloromethyl)propane
SMILES ClCC(CCl)(CCl)CCl

Specifications

Melting Point 96°C
Color White-Yellow
Boiling Point 110°C
Quantity 25 g
Formula Weight 209.92
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pentaerythrityl Tetrachloride