missing translation for 'onlineSavingsMsg'
Learn More

Pentaerythrityl Tetrabromide 98.0+%, TCI America™

Catalog No. p-7134750
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P07525G 5 g
P075225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P07525G Supplier TCI America Supplier No. P07525G
Only null left

Chemical Identifiers

CAS 3229-00-3
Molecular Formula C5H8Br4
Molecular Weight (g/mol) 387.74
MDL Number MFCD00000210
InChI Key OYSVBCSOQFXYHK-UHFFFAOYSA-N
Synonym 1,3-Dibromo-2,2-bis(bromomethyl)propane, Tetrakis(bromomethyl)methane
PubChem CID 76701
IUPAC Name 1,3-dibromo-2,2-bis(bromomethyl)propane
SMILES BrCC(CBr)(CBr)CBr

Specifications

Melting Point 161°C
Color White-Yellow
Boiling Point 306°C
Quantity 5 g
Formula Weight 387.74
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pentaerythrityl Tetrabromide
EINECSNumber (2)-0078
TSCA Yes