CAS RN 3229-00-3
CAS RN 3229-00-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Pentaerythrityl Tetrabromide 98.0+%, TCI America™
CAS: 3229-00-3 Molecular Formula: C5H8Br4 Molecular Weight (g/mol): 387.74 MDL Number: MFCD00000210 InChI Key: OYSVBCSOQFXYHK-UHFFFAOYSA-N Synonym: 1,3-Dibromo-2,2-bis(bromomethyl)propane, Tetrakis(bromomethyl)methane PubChem CID: 76701 IUPAC Name: 1,3-dibromo-2,2-bis(bromomethyl)propane SMILES: BrCC(CBr)(CBr)CBr