missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentaerythrityl Tetrabromide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 161°C |
| Color | White-Yellow |
| Boiling Point | 306°C |
| Quantity | 5 g |
| Formula Weight | 387.74 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Pentaerythrityl Tetrabromide |
Safety and Handling
| EINECSNumber | (2)-0078 |
| TSCA | Yes |