missing translation for 'onlineSavingsMsg'
Learn More

Pentaerythritol Tribromide 98.0+%, TCI America™

Supplier:  TCI America T1687500G

 View more versions of this product

Catalog No. T1687500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Pentaerythritol Tribromide
67°C
131°C
MFCD00021982
3-Bromo-2,2-bis(bromomethyl)-1-propanol, Tribromoneopentyl Alcohol, 2,2,2-Tris(bromomethyl)ethanol
C(C(CBr)(CBr)CBr)O
324.838
324.84
Crystalline Powder
1522-92-5
White
C5H9Br3O
500 g
QEJPOEGPNIVDMK-UHFFFAOYSA-N
3-bromo-2,2-bis(bromomethyl)propan-1-ol
15206
≥98.0% (T)

Chemical Identifiers

1522-92-5
324.838
QEJPOEGPNIVDMK-UHFFFAOYSA-N
15206
C(C(CBr)(CBr)CBr)O
C5H9Br3O
MFCD00021982
3-Bromo-2,2-bis(bromomethyl)-1-propanol, Tribromoneopentyl Alcohol, 2,2,2-Tris(bromomethyl)ethanol
3-bromo-2,2-bis(bromomethyl)propan-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2002

RTECSNumber : UA7410000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions