missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentaerythritol Tribromide 98.0+%, TCI America™
$65.46 - $286.85
Chemical Identifiers
| CAS | 1522-92-5 |
|---|---|
| Molecular Formula | C5H9Br3O |
| Molecular Weight (g/mol) | 324.838 |
| MDL Number | MFCD00021982 |
| InChI Key | QEJPOEGPNIVDMK-UHFFFAOYSA-N |
| Synonym | 3-Bromo-2,2-bis(bromomethyl)-1-propanol, Tribromoneopentyl Alcohol, 2,2,2-Tris(bromomethyl)ethanol |
| PubChem CID | 15206 |
| IUPAC Name | 3-bromo-2,2-bis(bromomethyl)propan-1-ol |
| SMILES | C(C(CBr)(CBr)CBr)O |
Chemical Identifiers
| 1522-92-5 | |
| 324.838 | |
| QEJPOEGPNIVDMK-UHFFFAOYSA-N | |
| 15206 | |
| C(C(CBr)(CBr)CBr)O |
| C5H9Br3O | |
| MFCD00021982 | |
| 3-Bromo-2,2-bis(bromomethyl)-1-propanol, Tribromoneopentyl Alcohol, 2,2,2-Tris(bromomethyl)ethanol | |
| 3-bromo-2,2-bis(bromomethyl)propan-1-ol |
Specifications
| 1522-92-5 | |
| White | |
| C5H9Br3O | |
| 25 g | |
| QEJPOEGPNIVDMK-UHFFFAOYSA-N | |
| 3-bromo-2,2-bis(bromomethyl)propan-1-ol | |
| 15206 | |
| ≥98.0% (T) | |
| Pentaerythritol Tribromide |
| 67°C | |
| 131°C | |
| MFCD00021982 | |
| 3-Bromo-2,2-bis(bromomethyl)-1-propanol, Tribromoneopentyl Alcohol, 2,2,2-Tris(bromomethyl)ethanol | |
| C(C(CBr)(CBr)CBr)O | |
| 324.838 | |
| 324.84 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-2002
RTECSNumber : UA7410000
TSCA : No