accessibility menu, dialog, popup

UserName

CAS RN 1522-92-5

OHBrBrBr

CAS RN 1522-92-5

IUPAC Name: 3-bromo-2,2-bis(bromomethyl)propan-1-ol
Synonyms: 3-bromo-2,2-bis(bromomethyl)propan-1-ol
Molecular Weight (g/mol): 324.84
Molecular Formula: C5H9Br3O
InChi Key: QEJPOEGPNIVDMK-UHFFFAOYSA-N
SMILES: OCC(CBr)(CBr)CBr
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results