missing translation for 'onlineSavingsMsg'
Learn More

Pentaerythritol Tetraacetate 98.0+%, TCI America™

Supplier:  TCI America P0757500G

 View more versions of this product

Catalog No. P0757500G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Pentaerythritol Tetraacetate
80°C
325°C
MFCD00026206
pentaerythritol tetraacetate, normosterol, normo-level, pentaerythrityl tetraacetate, pag romanian, tape, tetraacetyl pentestriol, tetraacetil pentoetriol, pentaerythritol, tetraacetate, unii-c4yxi01z81
CC(=O)OCC(COC(=O)C)(COC(=O)C)COC(=O)C
304.295
304.30
Crystalline Powder
597-71-7
White
C13H20O8
500 g
OUHCZCFQVONTOC-UHFFFAOYSA-N
[3-acetyloxy-2,2-bis(acetyloxymethyl)propyl] acetate
69007
≥98.0% (GC)

Chemical Identifiers

597-71-7
304.295
OUHCZCFQVONTOC-UHFFFAOYSA-N
69007
CC(=O)OCC(COC(=O)C)(COC(=O)C)COC(=O)C
C13H20O8
MFCD00026206
pentaerythritol tetraacetate, normosterol, normo-level, pentaerythrityl tetraacetate, pag romanian, tape, tetraacetyl pentestriol, tetraacetil pentoetriol, pentaerythritol, tetraacetate, unii-c4yxi01z81
[3-acetyloxy-2,2-bis(acetyloxymethyl)propyl] acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0642

RTECSNumber : RZ2600000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions