missing translation for 'onlineSavingsMsg'
Learn More

Pentaerythritol Tetraacetate 98.0+%, TCI America™

Catalog No. p-7134752
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P075725G 25 g
P0757500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P075725G Supplier TCI America Supplier No. P075725G

Chemical Identifiers

CAS 597-71-7
Molecular Formula C13H20O8
Molecular Weight (g/mol) 304.295
MDL Number MFCD00026206
InChI Key OUHCZCFQVONTOC-UHFFFAOYSA-N
Synonym pentaerythritol tetraacetate, normosterol, normo-level, pentaerythrityl tetraacetate, pag romanian, tape, tetraacetyl pentestriol, tetraacetil pentoetriol, pentaerythritol, tetraacetate, unii-c4yxi01z81
PubChem CID 69007
IUPAC Name [3-acetyloxy-2,2-bis(acetyloxymethyl)propyl] acetate
SMILES CC(=O)OCC(COC(=O)C)(COC(=O)C)COC(=O)C

Specifications

Melting Point 80°C
Color White
Boiling Point 325°C
Quantity 25 g
Formula Weight 304.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pentaerythritol Tetraacetate
EINECSNumber (2)-0642
RTECSNumber RZ2600000
TSCA Yes