accessibility menu, dialog, popup

UserName

CH3OOOH3COOCH3OOCH3O

CAS RN 597-71-7

IUPAC Name: 3-(acetyloxy)-2,2-bis[(acetyloxy)methyl]propyl acetate
Synonyms: pentaerythrityl tetraacetate
Molecular Weight (g/mol): 304.3
Molecular Formula: C13H20O8
InChi Key: OUHCZCFQVONTOC-UHFFFAOYSA-N
SMILES: CC(=O)OCC(COC(C)=O)(COC(C)=O)COC(C)=O
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results