missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentaerythritol Tetraacetate 98.0+%, TCI America™
Supplier: TCI America P075725G
Specifications
| Pentaerythritol Tetraacetate | |
| 80°C | |
| 325°C | |
| MFCD00026206 | |
| pentaerythritol tetraacetate, normosterol, normo-level, pentaerythrityl tetraacetate, pag romanian, tape, tetraacetyl pentestriol, tetraacetil pentoetriol, pentaerythritol, tetraacetate, unii-c4yxi01z81 | |
| CC(=O)OCC(COC(=O)C)(COC(=O)C)COC(=O)C | |
| 304.295 | |
| 304.30 | |
| Crystalline Powder |
| 597-71-7 | |
| White | |
| C13H20O8 | |
| 25 g | |
| OUHCZCFQVONTOC-UHFFFAOYSA-N | |
| [3-acetyloxy-2,2-bis(acetyloxymethyl)propyl] acetate | |
| 69007 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 597-71-7 | |
| 304.295 | |
| OUHCZCFQVONTOC-UHFFFAOYSA-N | |
| 69007 | |
| CC(=O)OCC(COC(=O)C)(COC(=O)C)COC(=O)C |
| C13H20O8 | |
| MFCD00026206 | |
| pentaerythritol tetraacetate, normosterol, normo-level, pentaerythrityl tetraacetate, pag romanian, tape, tetraacetyl pentestriol, tetraacetil pentoetriol, pentaerythritol, tetraacetate, unii-c4yxi01z81 | |
| [3-acetyloxy-2,2-bis(acetyloxymethyl)propyl] acetate |
Safety and Handling
EINECSNumber : (2)-0642
RTECSNumber : RZ2600000
TSCA : Yes