missing translation for 'onlineSavingsMsg'
Learn More

Pentadecane, ≥99.8% (GC), MilliporeSigma™ Supelco™

Pentadecane is a hydrocarbon, which is found to be highly viscous in nature. It is formed as a decomposition product of glycidyl palmitate of fatty acid, when subjected to heating in the temperature range of 180-200C.

$149.10 - $615.31

Specifications

CAS 629-62-9
Vapor Pressure 1 mmHg (91.6°C)
Vapor Density 7.4 (vs. air)
Density 0.769 g/mL (at 25°C)
Flash Point 132.00°C - Closed Cup
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111014949
View Documents
Merck Emd Millipore
765095ML
5 mL
Each for $149.10
Only null left
 
111014948
View Documents
Merck Emd Millipore
7650925ML
25 mL
Each for $615.31
Only null left
 
Description

Description

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications

Specifications

629-62-9
7.4 (vs. air)
132.00°C - Closed Cup
CH3(CH2)13 CH3
8°C to 10°C
Analytical Standard
≥99.8% (GC)
1 mmHg (91.6°C)
0.769 g/mL (at 25°C)
Neat
MFCD00008990
270°C
212.41
Analytical Standard
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'