missing translation for 'onlineSavingsMsg'
Learn More

Pentadecane, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014949
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014949 5 mL
111014948 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014949 Supplier Merck Emd Millipore Supplier No. 765095ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pentadecane is a hydrocarbon, which is found to be highly viscous in nature. It is formed as a decomposition product of glycidyl palmitate of fatty acid, when subjected to heating in the temperature range of 180-200C.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 629-62-9
Vapor Pressure 1 mmHg (91.6°C)
Vapor Density 7.4 (vs. air)
Density 0.769 g/mL (at 25°C)
Flash Point 132.00°C - Closed Cup
Physical Form Neat
Linear Formula CH3(CH2)13 CH3
MDL Number MFCD00008990
Melting Point 8°C to 10°C
Boiling Point 270°C
Type Analytical Standard
Quantity 5 mL
Formula Weight 212.41
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less