missing translation for 'onlineSavingsMsg'
Learn More

Pentachloronitrobenzene, Restek

Catalog No. 06719739
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06719739 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06719739 Supplier Chromatographic Supplier No. RK 32091

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 82-68-8
Molecular Formula C6Cl5NO2
Molecular Weight (g/mol) 295.321
InChI Key LKPLKUMXSAEKID-UHFFFAOYSA-N
PubChem CID 6720
ChEBI CHEBI:34908
IUPAC Name 1,2,3,4,5-pentachloro-6-nitrobenzene
SMILES C1(=C(C(=C(C(=C1Cl)Cl)Cl)Cl)Cl)[N+](=O)[O-]

Specifications

Quantity 1 mL
Chemical Name or Material Pentachloronitrobenzene