CAS RN 82-68-8
CAS RN 82-68-8
IUPAC Name:
1,2,3,4,5-pentachloro-6-nitrobenzene
Synonyms:
pentachloronitrobenzene PCNB quintozene nitropentachlorobenzene PKhNC
Molecular Weight (g/mol):
295.32
Molecular Formula:
C6Cl5NO2
InChi Key:
LKPLKUMXSAEKID-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=C(Cl)C(Cl)=C(Cl)C(Cl)=C1Cl
1
–
1
of
1
results
1
Pentachloronitrobenzene, Restek
CAS: 82-68-8 Molecular Formula: C6Cl5NO2 Molecular Weight (g/mol): 295.321 InChI Key: LKPLKUMXSAEKID-UHFFFAOYSA-N PubChem CID: 6720 ChEBI: CHEBI:34908 IUPAC Name: 1,2,3,4,5-pentachloro-6-nitrobenzene SMILES: C1(=C(C(=C(C(=C1Cl)Cl)Cl)Cl)Cl)[N+](=O)[O-]