Learn More
Pentacene, 97%
CAS: 135-48-8 | C22H14 | 278.35 g/mol
Supplier: Thermo Scientific Chemicals H5631003
Description
Pentacene can be used as OPV device (Organo Photovoltaic) and as semiconductors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPentacene can be used as OPV device (Organo Photovoltaic) and as semiconductors.
Solubility
Soluble in organic solvents.
Notes
Air and light sensitive. Store at cool temperature.
Specifications
| Pentacene | |
| 372°C to 374°C (sublimation) | |
| MFCD00003710 | |
| 1912418 | |
| 14,7107 | |
| Soluble in organic solvents. | |
| C1=CC2=CC3=CC4=CC5=CC=CC=C5C=C4C=C3C=C2C=C1 | |
| 278.35 | |
| CHEBI:33148 | |
| 97% |
| 135-48-8 | |
| C22H14 | |
| 1 g | |
| Air and light sensitive | |
| benzo b naphthacene, lin-dibenzanthracene, lin-naphthoanthracene, 2,3:6,7-dibenzanthracene, unii-9fqu5ha0uy, 9fqu5ha0uy, pentacen-2-yl, pentacene, 2,7-dibenzanthracene | |
| SLIUAWYAILUBJU-UHFFFAOYSA-N | |
| pentacene | |
| 8671 | |
| 278.35 |
Chemical Identifiers
| 135-48-8 | |
| 278.35 | |
| SLIUAWYAILUBJU-UHFFFAOYSA-N | |
| 8671 | |
| pentacene |
| C22H14 | |
| MFCD00003710 | |
| benzo b naphthacene, lin-dibenzanthracene, lin-naphthoanthracene, 2,3:6,7-dibenzanthracene, unii-9fqu5ha0uy, 9fqu5ha0uy, pentacen-2-yl, pentacene, 2,7-dibenzanthracene | |
| CHEBI:33148 | |
| C1=CC2=CC3=CC4=CC5=CC=CC=C5C=C4C=C3C=C2C=C1 |
Safety and Handling
EINECSNumber : 205-193-7
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only