missing translation for 'onlineSavingsMsg'
Learn More

Pentacene, 97%

Catalog No. p-7037550
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH56310MD 250 mg
AAH5631003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH56310MD Supplier Thermo Scientific Chemicals Supplier No. H56310MD
Only null left

CAS: 135-48-8 | C22H14 | 278.35 g/mol

Pentacene can be used as OPV device (Organo Photovoltaic) and as semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentacene can be used as OPV device (Organo Photovoltaic) and as semiconductors.

Solubility
Soluble in organic solvents.

Notes
Air and light sensitive. Store at cool temperature.

Chemical Identifiers

CAS 135-48-8
Molecular Formula C22H14
Molecular Weight (g/mol) 278.35
MDL Number MFCD00003710
InChI Key SLIUAWYAILUBJU-UHFFFAOYSA-N
Synonym benzo b naphthacene, lin-dibenzanthracene, lin-naphthoanthracene, 2,3:6,7-dibenzanthracene, unii-9fqu5ha0uy, 9fqu5ha0uy, pentacen-2-yl, pentacene, 2,7-dibenzanthracene
PubChem CID 8671
ChEBI CHEBI:33148
IUPAC Name pentacene
SMILES C1=CC2=CC3=CC4=CC5=CC=CC=C5C=C4C=C3C=C2C=C1

Specifications

Melting Point 372°C to 374°C (sublimation)
Quantity 250 mg
Beilstein 1912418
Sensitivity Air and light sensitive
Merck Index 14,7107
Solubility Information Soluble in organic solvents.
Formula Weight 278.35
Percent Purity 97%
Chemical Name or Material Pentacene
EINECSNumber 205-193-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only