missing translation for 'onlineSavingsMsg'
Learn More

Penicillin V potassium salt

Inhibits bacterial cell wall synthesis | CAS: 132-98-9 | C16H17KN2O5S | 388.48 g/mol

Supplier:  Thermo Scientific Chemicals J6244214

 View more versions of this product

Catalog No. AAJ6244214


Only null left
Explore available promotions

Description

Description

Penicillin V Potassium bactericidal against penicillin-susceptible microorganisms during the stage of active multiplication. It inhibits biosynthesis of cell-wall mucopeptide. It is used to treat or prevent infections that are proven or strongly suspected to be caused by bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Penicillin V Potassium bactericidal against penicillin-susceptible microorganisms during the stage of active multiplication. It inhibits biosynthesis of cell-wall mucopeptide. It is used to treat or prevent infections that are proven or strongly suspected to be caused by bacteria.

Solubility
Soluble in water, dimethyl sulfoxide, and methanol.

Notes
Store at -20°C. Incompatible with oxidizing agents. Protect from heat.
Specifications

Specifications

Penicillin V Potassium Salt
Powder
197°C to 202°C
25 g
MFCD00051771
14,7100
Soluble in water,dimethyl sulfoxide,and methanol.
[K+].CC1(C)S[C@@H]2[C@H](NC(=O)COC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
388.48
388.48
Penicillin V potassium salt
132-98-9
White
C16H17KN2O5S
3899451
Phenoxymethylpenicillinic acid potassium salt
HCTVWSOKIJULET-LQDWTQKMSA-M
(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenoxyacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;potassium
131673927

Chemical Identifiers

132-98-9
388.48
HCTVWSOKIJULET-LQDWTQKMSA-M
131673927
[K+].CC1(C)S[C@@H]2[C@H](NC(=O)COC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
C16H17KN2O5S
MFCD00051771
Phenoxymethylpenicillinic acid potassium salt
(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenoxyacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;potassium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
May cause an allergic skin reaction.

P261-P264b-P270-P272-P280g-P285-P301+P312-P302+P352-P304+P341-P330-P333+P313-P342+P311-P363-P501c

H302-H317-H334

EINECSNumber : 205-086-5

RTECSNumber : XH9275000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only