accessibility menu, dialog, popup

UserName

KCH3H3CS(R)(R)NHOOON(S)OO

CAS RN 132-98-9

IUPAC Name: potassium (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenoxyacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Synonyms: penicillin vk penicillin V potassium phenoxymethylpenicillin potassium
Molecular Weight (g/mol): 388.48
Molecular Formula: C16H17KN2O5S
InChi Key: HCTVWSOKIJULET-LQDWTQKMSA-M
SMILES: [K+].CC1(C)S[C@@H]2[C@H](NC(=O)COC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
Color 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Type 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results