missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Penciclovir
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 250 mg |
| Solubility | Solubility aq. soln.: 1.7 mg/mL (20°C). |
| Melting Point | 275.0°C to 277.0°C |
| Formula Weight | 253.26 |
| Chemical Name or Material | Penciclovir |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. |
RUO – Research Use Only