Learn More
Thermo Scientific Chemicals Pectinase, Aspergillus niger
Used in plant protoplast preparation to digest cell wall prior to organelle isolation. Pectinase helps facilitate the maceration of plant materials and extraction processes including organelle preparation.
Supplier:  Thermo Scientific Chemicals J6340803
 View more versions of this product
Catalog No. AAJ6340803
Only null left
Description
Description
Pectinase, Aspergillus niger is used in plant protoplast preparation to digest cell wall prior to organelle isolation. Pectinase helps facilitate the maceration of plant materials and extraction processes including organelle preparation. It is used in the enzyme preparation. Used mostly to reduce viscosity, to improve filtration to clarify products, to avoid particle sedimentation and to prevent pectin gel formation in the manufacture of fruit juice and wine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Chemical Identifiers
Powder
Specifications
1 g
Powder
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
EINECSNumber : 232-885-6
Recommended Storage : Ambient temperatures