missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD 153035 Hydrochloride 97.0+%, TCI America™
$132.35 - $365.35
Chemical Identifiers
| CAS | 183322-45-4 |
|---|---|
| Molecular Formula | C16H15BrClN3O2 |
| Molecular Weight (g/mol) | 396.669 |
| MDL Number | MFCD01074805 |
| InChI Key | ZJOKWAWPAPMNIM-UHFFFAOYSA-N |
| Synonym | pd153035 hydrochloride, pd 153035 hydrochloride, n-3-bromophenyl-6,7-dimethoxyquinazolin-4-amine hydrochloride, pd-153035 hydrochloride, tyrphostin ag 1517, ag 1517 hydrochloride, su-5271 hydrochloride, unii-ahj252p69n, 4-3-bromophenyl amino-6,7-dimethoxyquinazoline hydrochloride |
| PubChem CID | 11246488 |
| IUPAC Name | N-(3-bromophenyl)-6,7-dimethoxyquinazolin-4-amine;hydrochloride |
| SMILES | COC1=C(C=C2C(=C1)C(=NC=N2)NC3=CC(=CC=C3)Br)OC.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B494525MG
|
TCI America
B494525MG |
25 mg |
Each for $132.35
|
|
|||||
|
B4945100MG
|
TCI America
B4945100MG |
100 mg |
Each for $365.35
|
|
|||||
Chemical Identifiers
| 183322-45-4 | |
| 396.669 | |
| ZJOKWAWPAPMNIM-UHFFFAOYSA-N | |
| 11246488 | |
| COC1=C(C=C2C(=C1)C(=NC=N2)NC3=CC(=CC=C3)Br)OC.Cl |
| C16H15BrClN3O2 | |
| MFCD01074805 | |
| pd153035 hydrochloride, pd 153035 hydrochloride, n-3-bromophenyl-6,7-dimethoxyquinazolin-4-amine hydrochloride, pd-153035 hydrochloride, tyrphostin ag 1517, ag 1517 hydrochloride, su-5271 hydrochloride, unii-ahj252p69n, 4-3-bromophenyl amino-6,7-dimethoxyquinazoline hydrochloride | |
| N-(3-bromophenyl)-6,7-dimethoxyquinazolin-4-amine;hydrochloride |
Specifications
| 183322-45-4 | |
| C16H15BrClN3O2 | |
| 25 mg | |
| ZJOKWAWPAPMNIM-UHFFFAOYSA-N | |
| N-(3-bromophenyl)-6,7-dimethoxyquinazolin-4-amine;hydrochloride | |
| 11246488 | |
| ≥97.0% (HPLC) | |
| PD 153035 Hydrochloride |
| 265°C | |
| MFCD01074805 | |
| pd153035 hydrochloride, pd 153035 hydrochloride, n-3-bromophenyl-6,7-dimethoxyquinazolin-4-amine hydrochloride, pd-153035 hydrochloride, tyrphostin ag 1517, ag 1517 hydrochloride, su-5271 hydrochloride, unii-ahj252p69n, 4-3-bromophenyl amino-6,7-dimethoxyquinazoline hydrochloride | |
| COC1=C(C=C2C(=C1)C(=NC=N2)NC3=CC(=CC=C3)Br)OC.Cl | |
| 396.669 | |
| 396.67 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer (-20 °C)