missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD 153035 Hydrochloride 97.0+%, TCI America™
Supplier: TCI America B4945100MG
Specifications
| PD 153035 Hydrochloride | |
| 265°C | |
| MFCD01074805 | |
| pd153035 hydrochloride, pd 153035 hydrochloride, n-3-bromophenyl-6,7-dimethoxyquinazolin-4-amine hydrochloride, pd-153035 hydrochloride, tyrphostin ag 1517, ag 1517 hydrochloride, su-5271 hydrochloride, unii-ahj252p69n, 4-3-bromophenyl amino-6,7-dimethoxyquinazoline hydrochloride | |
| COC1=C(C=C2C(=C1)C(=NC=N2)NC3=CC(=CC=C3)Br)OC.Cl | |
| 396.669 | |
| 396.67 | |
| Crystalline Powder |
| 183322-45-4 | |
| C16H15BrClN3O2 | |
| 100 mg | |
| ZJOKWAWPAPMNIM-UHFFFAOYSA-N | |
| N-(3-bromophenyl)-6,7-dimethoxyquinazolin-4-amine;hydrochloride | |
| 11246488 | |
| ≥97.0% (HPLC) |
Chemical Identifiers
| 183322-45-4 | |
| 396.669 | |
| ZJOKWAWPAPMNIM-UHFFFAOYSA-N | |
| 11246488 | |
| COC1=C(C=C2C(=C1)C(=NC=N2)NC3=CC(=CC=C3)Br)OC.Cl |
| C16H15BrClN3O2 | |
| MFCD01074805 | |
| pd153035 hydrochloride, pd 153035 hydrochloride, n-3-bromophenyl-6,7-dimethoxyquinazolin-4-amine hydrochloride, pd-153035 hydrochloride, tyrphostin ag 1517, ag 1517 hydrochloride, su-5271 hydrochloride, unii-ahj252p69n, 4-3-bromophenyl amino-6,7-dimethoxyquinazoline hydrochloride | |
| N-(3-bromophenyl)-6,7-dimethoxyquinazolin-4-amine;hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Freezer (-20 °C)