missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PCR Master Mix Starter Pack

Catalog No. A31021
Encompass
Change view
Click to view available options
Quantity:
1 pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31021 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A31021 Supplier Invitrogen™ Supplier No. A31021

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The PCR Master Mix Starter Pack is composed of three different PCR master mixes for easy evaluation of their suitability for use in your research.

The PCR Master Mix Starter Pack is composed of three different PCR master mixes for easy evaluation of their suitability for use in your research. These master mixes provide our best DNA polymerases for routine PCR, hot-start PCR, and high-fidelity PCR needs. Thermo Scientific DreamTaq Green PCR Master Mix contains an enhanced Taq DNA polymerase optimized for all standard PCR applications. Invitrogen Platinum II Hot Start PCR Master Mix contains hot-start Taq DNA polymerase for specific, robust, and reliable PCR. Invitrogen SuperFi II PCR Master Mix features superior performance and an error rate 300 times lower than that of Taq. It is an excellent choice for cloning, mutagenesis, and other applications requiring high fidelity.

Master mixes included in this starter pack:

  • 2X DreamTaq Green PCR Master Mix (2X), 200 reactions (Cat. No. K1081)
  • 2X Platinum II Hot Start PCR Master Mix (2X), 50 reactions (14000012)
  • Platinum SuperFi II PCR Master Mix, 100 reactions (12368010)

Specifications

Content And Storage • 2X DreamTaq Green PCR Master Mix, 200 reactions
• 2X Platinum II Hot-Start PCR Master Mix, 50 reactions
• 2X Platinum SuperFi II PCR Master Mix, 100 reactions

Store all components at -20°C.
Polymerase Platinum II Taq Hot Start DNA Polymerase
Kit Type PCR Master Mix Starter Pack
Product Type PCR Master Mix Starter Pack
Quantity 1 pack
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR
No. of Reactions 350 Reactions
Reaction Format Kit
Starting Material DNA

For Research Use Only. Not for use in diagnostic procedures.