missing translation for 'onlineSavingsMsg'
Learn More

Promega PCR Master Mix

Catalog No. p-2902701
Encompass
Change view
Click to view available options
No. of Reactions:
100 x 50& mu;L Reactions
1000 x 50 μL Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
PRM7505 1000 x 50 μL Reactions
PRM7502 100 x 50& mu;L Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRM7505 Supplier Promega Supplier No. M7505
Only null left

Includes

PCR Master Mix, Nuclease-free water

Premixed 2X solution of Taq DNA Polymerase, dNTPs and Reaction Buffer.

  • Premixed 2X solution of Taq DNA Polymerase, dNTPs and Reaction Buffer
  • One pipetting step adds Taq DNA polymerase, dNTPs, MgCl2 and reaction buffer
  • Set up reactions with as low as 10 μL volume

Specifications

Concentration 2X
Content And Storage • 2X PCR Master Mix, 1 x 25 mL
• Nuclease-free Water, 1 x 25 mL
• Taq DNA Polymerase
• dNTPs
• MgCl2
• Reaction Buffers

Store at –30° to –10°C.
For Use With (Application) Standard PCR, RT-PCR
No. of Reactions 1000 x 50 μL Reactions
Volume 50 μL