missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega PCR Master Mix
Description
Includes
PCR Master Mix, Nuclease-free water
- Premixed 2X solution of Taq DNA Polymerase, dNTPs and Reaction Buffer
- One pipetting step adds Taq DNA polymerase, dNTPs, MgCl2 and reaction buffer
- Set up reactions with as low as 10 μL volume
Specifications
Specifications
| Concentration | 2X |
| Content And Storage | • 2X PCR Master Mix, 1 x 25 mL • Nuclease-free Water, 1 x 25 mL • Taq DNA Polymerase • dNTPs • MgCl2 • Reaction Buffers Store at –30° to –10°C. |
| For Use With (Application) | Standard PCR, RT-PCR |
| No. of Reactions | 1000 x 50 μL Reactions |
| Volume | 50 μL |