missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PBS, pH 7.4 (flow cytometry grade)

Catalog No. A1286301
Change view
Click to view available options
Quantity:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1286301 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1286301 Supplier Gibco™ Supplier No. A1286301

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Phosphate-Buffered Saline (PBS) is a balanced salt solution used for a variety of applications, including reagent preparation, diluting cells for flow cytometry and as a cell culture reagent.

Phosphate-Buffered Saline (PBS) is a balanced salt solution used for a variety of applications, including reagent preparation, diluting cells for flow cytometry and as a cell culture reagent. This Gibco™ PBS is formulated without calcium and magnesium for suspending cells for flow cytometry. This PBS is not intended for cell culture as it is not tested for sterility. We offer a variety of Gibco™ PBS formulations for cell culture applications. Find the right cell culture product using the media selector tool.

This PBS is manufactured as follows:
Without: Calcium, Magnesium, Phenol Red

Product Intended Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Concentration 1 X
pH 7.4
Physical Form Liquid
Quantity 10 L
Solution Type Phosphate Buffered Saline
Sterility Sterile-filtered
Includes No Calcium, No Magnesium
Manufacturing Quality cGMP-compliant under the ISO 13485 standard

For Research Use Only. Not for use in diagnostic procedures.