missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PBS, pH 7.4

Catalog No. p-200110071
Change view
Click to view available options
Quantity:
5 L
20 L
50 L
100 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A58313BA 5 L
A58313BB 20 L
A58313BD 50 L
A58313BE 100 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A58313BA Supplier Gibco™ Supplier No. A58313BA
Only null left

Gibco™ Balanced Salt Solutions are sterile-filtered, packaged in bioprocess containers, and ready to order.

Gibco™ Balanced Salt Solutions are sterile-filtered, packaged in bioprocess containers, and ready to order. Commonly used for processing cells and flushing equipment, sourcing Gibco balanced salt solutions can help streamline your operation by reducing buffer preparation and allowing you to focus on additional critical steps in your biopharmaceutical processes. Discover new outsourcing efficiencies to reclaim time, space, and resources.

Product Features

  • Multiple formulations and concentrations available
  • Available in a range of pack sizes (5 L, 20 L, 50 L, 100 L, and 200 L)
  • Established products with set specifications are made to order
  • Product standardization enables shorter times from inquiry to quote and shipment, compared to custom
  • Readily accessible documentation for accelerated qualification

Benefits

  • Reduces time spent preparing buffers in-house
  • Mitigates space requirement for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Meets workflow needs across stages of development and manufacturing of biologicals

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Color Clear, colorless solution
pH 7.3 to 7.5
Physical Form Liquid
Quantity 5 L
Sterility Sterile-filtered
Includes Calcium, Magnesium, Phenol Red
Manufacturing Quality cGMP
Osmolality 280 to 315 mOsm/kg

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.