missing translation for 'onlineSavingsMsg'
Learn More

PBS, Phosphate Buffered Saline, 1X Powder, pH 7.4, Fisher BioReagents

Catalog No. BP66150
Change view
Click to view available options
Quantity:
10 L
50 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP66150 50 L
BP66110 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP66150 Supplier Fisher BioReagents Supplier No. FLBP661 50
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

  • Provides a convenient way to make 50L of standard 1X phosphate buffer
  • Reconstitute contents of bottle in ultrapure water, such as BP2819, to yield a 1X PBS solution containing 0.137M NaCl, 0.0027M KCl, and 0.0119M phosphates
  • Tested for the absence of DNase, RNase, and protease
  • PBS is a standard buffer used in many biochemical applications to maintain pH
  • Intended for laboratory research use only

Specifications

Color White
Physical Form Powder
Quantity 50 L
Packaging Poly Bottle