missing translation for 'onlineSavingsMsg'
Learn More

Phosphate Buffered Saline (PBS), 10X Powder, pH 7.4, Fisher BioReagents™

Catalog No. BP6651
Change view
Click to view available options
Quantity:
2 x 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP6651 2 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP6651 Supplier Fisher BioReagents Supplier No. FLBP665 1
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Phosphate Buffered Saline (PBS), 10X Powder provides a convenient way to make 1 L of 10X stock PBS solution.

  • Provides a convenient way to make 1 L of 10X stock PBS solution
  • Eliminates the need to weigh and mix individual components
  • When reconstituted in ultrapure water such as BP2819, each packet yields 1 L of a 10X PBS solution containing 1.37M NaCl, 0.027M KCl, and 0.119M phosphates
  • Tested for the absence of DNase, RNase, and protease
  • PBS is a standard buffer used in many biochemical applications to maintain pH
  • Intended for laboratory research use only

Specifications

Color White
Physical Form Solid
Quantity 2 x 1 L
Packaging Foil Pack