missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Dulbecco's Phosphate Buffered Saline (10X) without Calcium and Magnesium

Catalog No. ICN1960454
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN1960454 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN1960454 Supplier MP Biomedicals™ Supplier No. 091960454

Stock solution used to prepare 1X D-PBS in cell culture grade water.

Dulbecco's Phosphate Buffered Saline 10X is a stock solution used to prepare 1X D-PBS in cell culture grade water. D-PBS is used to wash cells during preparation and serial transfer. Phosphate buffering maintains the pH in the physiological range. Calcium and magnesium facilitate cell binding and clumping. DPBS without these ions can be used to wash and rinse suspended cells. Used to wash cells during preparation and serial transfer.

Specifications

Chemical Name or Material DPBS
Concentration 10 X
pH 6.5 to 7.5
Physical Form Solution
Quantity 500 mL
Formulation Liquid (as 10X) Inorganic Salts: Potassium Chloride [KCl]: 2000.00 mg/L; Potassium Phosphate Monobasic [KH2PO4]: 2000.00 mg/L; Sodium Chloride [NaCl]: 80000.00 mg/L; Sodium Phosphate Dibasic [Na2HPO4]: 11500.00 mg/L
Sterility Sterile
Includes No Calcium, No Magnesium

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.