Learn More
Thermo Scientific Chemicals Patent Blue VF, pure, indicator grade
CAS: 129-17-9 | C27H31N2NaO6S2 | 566.66 g/mol
$101.78 - $372.51
Chemical Identifiers
| CAS | 129-17-9 |
|---|---|
| Molecular Formula | C27H31N2NaO6S2 |
| Molecular Weight (g/mol) | 566.66 |
| MDL Number | MFCD00012119 |
| InChI Key | SJEYSFABYSGQBG-UHFFFAOYSA-M |
| Synonym | Acid Blue 1, C.I. 42045, αzurine 2G, Sulphan blue |
| PubChem CID | 8507 |
| ChEBI | CHEBI:34906 |
| IUPAC Name | sodium;4-[[4-(diethylamino)phenyl]-(4-diethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]benzene-1,3-disulfonate |
| SMILES | [Na+].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1S([O-])(=O)=O)S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC189410250
|
Thermo Scientific Chemicals
189410250 |
25 g |
Each for $101.78
|
|
|||||
|
AC189411000
|
Thermo Scientific Chemicals
189411000 |
100 g |
Each for $372.51
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
| 129-17-9 | |
| 566.66 | |
| SJEYSFABYSGQBG-UHFFFAOYSA-M | |
| 8507 | |
| sodium;4-[[4-(diethylamino)phenyl]-(4-diethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]benzene-1,3-disulfonate |
| C27H31N2NaO6S2 | |
| MFCD00012119 | |
| Acid Blue 1, C.I. 42045, αzurine 2G, Sulphan blue | |
| CHEBI:34906 | |
| [Na+].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1S([O-])(=O)=O)S([O-])(=O)=O |
Specifications
| 129-17-9 | |
| MFCD00012119 | |
| Solubility in water: 50g/L (20°C). Other solubilities: 40g/L (95°C), 30 mg/l in egme, 7 mg/l in etoh | |
| Authentic | |
| sodium;4-[[4-(diethylamino)phenyl]-(4-diethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]benzene-1,3-disulfonate | |
| 8507 | |
| 566.67 | |
| Glass bottle | |
| Complies | |
| 25 g | |
| Patent Blue VF |
| C27H31N2NaO6S2 | |
| Acid Blue 1, C.I. 42045, αzurine 2G, Sulphan blue | |
| SJEYSFABYSGQBG-UHFFFAOYSA-M | |
| [Na+].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1S([O-])(=O)=O)S([O-])(=O)=O | |
| 566.66 | |
| CHEBI:34906 | |
| Pure | |
| suitable as acid-base indicator | |
| Blue/Green to Purple | |
| Crystalline Powder |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CANCER SUSPECTED AGENT
EINECSNumber : 204-934-1
RTECSNumber : BP6830000
TSCA : TSCA
RUO – Research Use Only