missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Patent Blue VF, pure, indicator grade

CAS: 129-17-9 | C27H31N2NaO6S2 | 566.66 g/mol

Supplier:  Thermo Scientific Chemicals 189410250

 View more versions of this product

Catalog No. AC189410250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

Patent Blue VF
C27H31N2NaO6S2
Acid Blue 1, C.I. 42045, αzurine 2G, Sulphan blue
SJEYSFABYSGQBG-UHFFFAOYSA-M
[Na+].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1S([O-])(=O)=O)S([O-])(=O)=O
566.66
CHEBI:34906
Pure
suitable as acid-base indicator
Blue/Green to Purple
Crystalline Powder
129-17-9
MFCD00012119
Solubility in water: 50g/L (20°C). Other solubilities: 40g/L (95°C), 30 mg/l in egme, 7 mg/l in etoh
Authentic
sodium;4-[[4-(diethylamino)phenyl]-(4-diethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]benzene-1,3-disulfonate
8507
566.67
Glass bottle
Complies
25 g

Chemical Identifiers

129-17-9
566.66
SJEYSFABYSGQBG-UHFFFAOYSA-M
8507
sodium;4-[[4-(diethylamino)phenyl]-(4-diethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]benzene-1,3-disulfonate
C27H31N2NaO6S2
MFCD00012119
Acid Blue 1, C.I. 42045, αzurine 2G, Sulphan blue
CHEBI:34906
[Na+].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1S([O-])(=O)=O)S([O-])(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

CANCER SUSPECTED AGENT

EINECSNumber : 204-934-1

RTECSNumber : BP6830000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only