accessibility menu, dialog, popup

UserName

NaCH3NCH3CNCH3H3CSOOOSOOO

CAS RN 129-17-9

IUPAC Name: sodium bis[4-(diethylamino)phenyl](2,4-disulfonatophenyl)methylium
Synonyms: sodium bis[4-(diethylamino)phenyl](2,4-disulfonatophenyl)methylium
Molecular Weight (g/mol): 566.66
Molecular Formula: C27H31N2NaO6S2
InChi Key: SJEYSFABYSGQBG-UHFFFAOYSA-M
SMILES: [Na+].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1S([O-])(=O)=O)S([O-])(=O)=O
Color 
  • (3)
  • (2)
Grade 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)