Learn More
Thermo Scientific™ Particle Certified HDPE Containers
GREENER_CHOICE
Protect the quality of your products or lab samples with these reliable particle-certified HDPE containers,certified to contain < 20 particles per milliliter = 0.3 microns.
$717.03 - $1,445.41
Specifications
Color Translucent
Material HDPE
Product Line Nalgene
Certifications/Compliance Low metals certification includes aluminum, calcium, copper, iron, potassium, magnesium, manganese, sodium, and zinc at less than 10 ppb. A certificate of analysis is included with each lot.
Closure Included Yes
View More Specs
Products 6
Catalog Number Mfr. No. Capacity (Metric) Certified Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Certified Price Quantity & Availability  
12200118
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
156 125W/N/LPM
125 mL Low Particulate, Low Metals
Case of 24 Each for $1,445.41
Only null left
12200117
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
156 125W/N/LP
125 mL Low Particulate
Case of 24 Each for $721.56
Only null left
12200119
View Documents Promotion Details
Thermo Scientific™
157 250W/N/LPM
250 mL Low Particulate, Low Metals
Case of 24 Each for $1,350.09
Only null left
12200140
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
157 250W/N/LP
250 mL Low Particulate
Case of 24 Each for $1,142.82
Only null left
12200116
View Documents Promotion Details
Thermo Scientific™
150 01W/N/LPM
1000 mL Low Particulate, Low Metals
Case of 12 Each for $1,007.47
Only null left
12200139
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
150 01W/N/LP
1000 mL Low Particulate
Case of 12 Each for $717.03
Only null left
Description
Description
Protect the quality of your products or lab samples with reliable Thermo Scientific™ Particle-Certified HDPE Containers, tested and certified to contain <20 particles per milliliter = 0.3 microns. A Certificate of Analysis is included with each shipment.
A large variety of standard particle-certified containers are available. Nonstandard containers may be custom cleaned on request.
Key Features:
  • Narrow-mouth
  • Includes leak-proof† polypropylene caps
  • Suitable for use in sampling and packaging, including combination packaging for shipping of hazardous materials
  • Double-bagged and ready for cleanroom with no extra preparation
Recommended for:
Low particle applications
Specifications
Specifications
Translucent
Nalgene
Yes
Round
Upstream process solution storage. Upstream Validation. Upstream process solution storage
HDPE
Low metals certification includes aluminum, calcium, copper, iron, potassium, magnesium, manganese, sodium, and zinc at less than 10 ppb. A certificate of analysis is included with each lot.
Narrow
Polypropylene
Videos
SDS
Documents
Documents
†The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.