missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Particle Certified HDPE Containers

Protect the quality of your products or lab samples with these reliable particle-certified HDPE containers,certified to contain < 20 particles per milliliter = 0.3 microns.

Supplier:  Thermo Scientific™ 150 01W/N/LPM

 View more versions of this product

Catalog No. 12200116


Only null left

Description

Description

Protect the quality of your products or lab samples with reliable Thermo Scientific™ Particle-Certified HDPE Containers, tested and certified to contain <20 particles per milliliter = 0.3 microns. A Certificate of Analysis is included with each shipment.

A large variety of standard particle-certified containers are available. Nonstandard containers may be custom cleaned on request.

Key Features:

  • Narrow-mouth
  • Includes leak-proof† polypropylene caps
  • Suitable for use in sampling and packaging, including combination packaging for shipping of hazardous materials
  • Double-bagged and ready for cleanroom with no extra preparation

Recommended for:

Low particle applications

Specifications

Chemical Identifiers

Translucent
Nalgene
Yes
Round
Upstream process solution storage. Upstream Validation. Upstream process solution storage
HDPE
Low metals certification includes aluminum, calcium, copper, iron, potassium, magnesium, manganese, sodium, and zinc at less than 10 ppb. A certificate of analysis is included with each lot.
Narrow
Polypropylene

Specifications

32 oz., 1000mL
32 oz.
3.625 in.
12
Low Particulate, Low Metals
1000mL
Yes
Round
Square Water Sampling Bottle
Polypropylene
92.08 mm
1000 mL
Translucent
HDPE
Nalgene
32 oz.
Low metals certification includes aluminum, calcium, copper, iron, potassium, magnesium, manganese, sodium, and zinc at less than 10 ppb. A certificate of analysis is included with each lot.
Narrow
215.9 mm
8.5 in.
38-430 mm
Upstream process solution storage. Upstream Validation. Upstream process solution storage
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

†The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.