missing translation for 'onlineSavingsMsg'
Learn More

Pararosaniline 97.0+%, TCI America™

Catalog No. p-7134380
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P002125G 25 g
P0021500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P002125G Supplier TCI America Supplier No. P002125G
Only null left

Chemical Identifiers

CAS 467-62-9
Molecular Formula C19H19N3O
Molecular Weight (g/mol) 305.381
MDL Number MFCD00036222
InChI Key KRVRUAYUNOQMOV-UHFFFAOYSA-N
Synonym pararosaniline base, tris 4-aminophenyl methanol, tris p-aminophenyl methanol, benzenemethanol, 4-amino-.alpha.,.alpha.-bis 4-aminophenyl, tris 4-aminophenyl methan-1-ol, fuchsin dye base, parafuchsin carbinol, tris 4-aminophenyl carbinol, methanol, tris 4-aminophenyl, 4,4',4-triaminotrityl alcohol
PubChem CID 10084
IUPAC Name tris(4-aminophenyl)methanol
SMILES C1=CC(=CC=C1C(C2=CC=C(C=C2)N)(C3=CC=C(C=C3)N)O)N

Specifications

Melting Point 205°C
Color Purple-Red
Quantity 25 g
Formula Weight 305.38
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pararosaniline