accessibility menu, dialog, popup

UserName

CAS RN 467-62-9

IUPAC Name: tris(4-aminophenyl)methanol
Synonyms: tris(4-aminophenyl)methanol
Molecular Weight (g/mol): 305.38
Molecular Formula: C19H19N3O
InChi Key: KRVRUAYUNOQMOV-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C(O)(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)