missing translation for 'onlineSavingsMsg'
Learn More

Pararosaniline Hydrochloride 95.0+%, TCI America™

Supplier:  TCI America P205225G

 View more versions of this product

Catalog No. P205225G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Pararosaniline Hydrochloride [for Biochemical Research]
270°C
C19H18ClN3
25 g
JUQPZRLQQYSMEQ-UHFFFAOYSA-N
4-[(4-aminophenyl)-(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride
11292
323.82
Crystalline Powder
569-61-9
Black-Green
MFCD00001657
basic red 9, pararosaniline hydrochloride, parafuchsin, pararosaniline chloride, parafuchsine, p-fuchsin, basic fuchsin, ci basic red 9, c.i. basic red 9, parafuksin
C1=CC(=N)C=CC1=C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N.Cl
323.824
CHEBI:87663
≥95.0% (N)

Chemical Identifiers

569-61-9
323.824
JUQPZRLQQYSMEQ-UHFFFAOYSA-N
11292
4-[(4-aminophenyl)-(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride
C19H18ClN3
MFCD00001657
basic red 9, pararosaniline hydrochloride, parafuchsin, pararosaniline chloride, parafuchsine, p-fuchsin, basic fuchsin, ci basic red 9, c.i. basic red 9, parafuksin
CHEBI:87663
C1=CC(=N)C=CC1=C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N.Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : CX9850100

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions