missing translation for 'onlineSavingsMsg'
Learn More

Pararosaniline Hydrochloride 95.0+%, TCI America™

Catalog No. p-7135380
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
P20525G 5 g
P205225G 25 g
2 options

Catalog No. P20525G

Supplier: TCI America P20525G

Only null left

Chemical Identifiers

CAS 569-61-9
Molecular Formula C19H18ClN3
Molecular Weight (g/mol) 323.824
MDL Number MFCD00001657
InChI Key JUQPZRLQQYSMEQ-UHFFFAOYSA-N
Synonym basic red 9, pararosaniline hydrochloride, parafuchsin, pararosaniline chloride, parafuchsine, p-fuchsin, basic fuchsin, ci basic red 9, c.i. basic red 9, parafuksin
PubChem CID 11292
ChEBI CHEBI:87663
IUPAC Name 4-[(4-aminophenyl)-(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride
SMILES C1=CC(=N)C=CC1=C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N.Cl

Specifications

Melting Point 270°C
Color Black-Green
Quantity 5 g
Formula Weight 323.82
Percent Purity ≥95.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Pararosaniline Hydrochloride [for Biochemical Research]
RTECSNumber CX9850100
TSCA Yes