missing translation for 'onlineSavingsMsg'
Learn More

Pararosaniline base

Catalog No. p-178309
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC201550100 10 g Glass bottle
AC201550500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC201550100 Supplier Thermo Scientific Chemicals Supplier No. 201550100
Only null left

CAS: 467-62-9 | C19H19N3O | 305.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 467-62-9
Molecular Formula C19H19N3O
Molecular Weight (g/mol) 305.38
MDL Number MFCD00036222
InChI Key KRVRUAYUNOQMOV-UHFFFAOYSA-N
Synonym pararosaniline base, tris 4-aminophenyl methanol, tris p-aminophenyl methanol, benzenemethanol, 4-amino-.alpha.,.alpha.-bis 4-aminophenyl, tris 4-aminophenyl methan-1-ol, fuchsin dye base, parafuchsin carbinol, tris 4-aminophenyl carbinol, methanol, tris 4-aminophenyl, 4,4',4-triaminotrityl alcohol
PubChem CID 10084
IUPAC Name tris(4-aminophenyl)methanol
SMILES C1=CC(=CC=C1C(C2=CC=C(C=C2)N)(C3=CC=C(C=C3)N)O)N

Specifications

Melting Point 205.0°C
Color Purple
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 10 g
Beilstein 13, 733
Solubility Information Solubility in water: very slightly soluble. Other solubilities: soluble in alcohol,slightly soluble in ether
Formula Weight 305.38
Physical Form Crystalline Powder
Chemical Name or Material Pararosaniline base
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of causing cancer.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
EINECSNumber 207-395-

RUO – Research Use Only